Assay Details
Gene Symbol: Polr3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012081.1 | NP_001012081.1 | 14 - 15 | 1887 | 92 | |
XM_008761310.1 | XP_008759532.1 | 14 - 15 | 1817 | 92 | ||
XM_008761311.1 | XP_008759533.1 | 13 - 14 | 2044 | 92 | ||
XM_008761312.1 | XP_008759534.1 | 13 - 14 | 1908 | 92 | ||
GenBank mRNA | BC083667.1 | - | 14 - 15 | 1887 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
