Assay Details
Gene Symbol: Rbm8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271138.1 | NP_001258067.1 | 3 - 4 | 251 | 63 | |
XM_006232956.2 | XP_006233018.1 | 3 - 4 | 249 | 63 | ||
GenBank mRNA | DQ359102.1 | - | 3 - 4 | 203 | 63 | |
FQ210627.1 | - | 3 - 4 | 224 | 63 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
