Assay Details
Gene Symbol: Ctsk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031560.2 | NP_113748.1 | 1 - 2 | 74 | 86 | |
GenBank mRNA | BC078793.1 | - | 1 - 2 | 74 | 86 | |
FQ228691.1 | - | 1 - 2 | 91 | 86 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC078793.1, NM_031560.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
