Assay Details
Gene Symbol: Pdzk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031712.1 | NP_113900.1 | 5 - 6 | 861 | 87 | |
GenBank mRNA | AF116896.1 | - | 5 - 6 | 861 | 87 | |
BC078788.1 | - | 5 - 6 | 853 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
