Assay Details
Gene Symbol: Ecm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053882.1 | NP_446334.1 | 5 - 6 | 511 | 71 | |
XM_006232942.2 | XP_006233004.1 | 5 - 6 | 843 | 71 | ||
GenBank mRNA | BC085879.1 | - | 5 - 6 | 511 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
