Assay Details
Gene Symbol: Polr3gl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109571.1 | NP_001103041.1 | 4 - 5 | 445 | 71 | |
XM_008761331.1 | XP_008759553.1 | 4 - 5 | 528 | 71 | ||
GenBank mRNA | BC168229.1 | - | 4 - 5 | 404 | 71 | |
FQ214367.1 | - | 4 - 5 | 431 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
