Assay Details
Gene Symbol: Polr3gl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109571.1 | NP_001103041.1 | 7 - 8 | 682 | 155 | |
GenBank mRNA | BC168229.1 | - | 7 - 8 | 641 | 155 | |
FQ214367.1 | - | 7 - 8 | 668 | 155 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
