Assay Details
Gene Symbol: Casq2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017131.2 | NP_058827.3 | 1 - 2 | 383 | 139 | |
GenBank mRNA | BC072547.1 | - | 1 - 2 | 370 | 139 | |
CK602439.1 | - | 1 - 2 | 383 | 139 | ||
U33287.1 | - | 1 - 2 | 364 | 139 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
