Assay Details
Gene Symbol: Igsf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106455.1 | NP_001099925.1 | 5 - 6 | 1651 | 88 | |
XM_006233041.2 | XP_006233103.1 | 7 - 8 | 2454 | 88 | ||
XM_006233042.2 | XP_006233104.1 | 7 - 8 | 2451 | 88 | ||
XM_006233043.2 | XP_006233105.1 | 6 - 7 | 2394 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
