Assay Details
Gene Symbol: Cdc42se1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039044.1 | NP_001034133.1 | 3 - 4 | 439 | 91 | |
GenBank mRNA | BC091418.1 | - | 3 - 4 | 439 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
