Assay Details
Gene Symbol: Man1a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106452.1 | NP_001099922.1 | 12 - 13 | 2150 | 110 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P078592 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 313303 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 310589 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 145369 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 558022 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 657066 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 931351 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 24159 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 570611 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 189595 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 38929 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 423661 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 141627 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 157622 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 304815 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427779 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 685552 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 213568 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 932590 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 389764 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 296306 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 890775 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 632938 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 424043 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 643319 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 717635 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 576944 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 950553 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881591 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 350997 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 526177 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 511771 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 956537 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 177406 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 58179 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 286680 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 718847 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 122333 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 455726 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 65627 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 346865 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 970751 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 794899 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 361611 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 34644 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 454306 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 964944 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 784388 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 80512 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 343333 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 432117 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 706153 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 174321 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 643155 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 116677 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 964712 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 808372 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 684959 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 340107 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 557052 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 921096 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 577990 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 535067 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 9884 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 84504 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 472750 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 683767 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 475568 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 622430 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 472111 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 69333 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 734411 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 330333 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 659474 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 389622 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 49943 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 133752 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 613482 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 445734 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 505153 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 209289 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 375611 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 668730 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 290223 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 627173 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 821152 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 441622 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 172450 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 879210 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 302362 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 899085 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 596753 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 774692 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 421657 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 822644 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 184883 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 108801 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 175347 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 148002 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 168652 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 2119 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 91313 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 5391 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 148814 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 85623 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 40977 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 55642 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 76703 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 81968 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 46474 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P078592 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P078592 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P078592 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P450173 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 36390 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 65800 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10825407 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10825411 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10825417 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10825409 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10825413 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10825415 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10825414 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10825412 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10825410 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10825416 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10825408 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10825406 | - | - |
Rat Genome 230 2.0 Array | 1394346_PM_at | - | - |
Rat Genome 230 2.0 Array | 1375261_at | - | - |
Rat Genome 230 2.0 Array | 1394346_at | - | - |
Rat Genome 230 2.0 Array | 1375261_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0200029671.rn.1 | TC0200004317.rn.1 | RTA TC |
Rat Expression Set 230 | 1375261_at | - | - |
Rat Expression Set 230 | 1375261_PM_at | - | - |
Rat Expression Set 230 | 1394346_at | - | - |
Rat Expression Set 230 | 1394346_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10825406 | - | - |
Rat Genome U34 Set | rc_AI011360_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21673478 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1650898 | - | - |
