Assay Details
Gene Symbol: Gdap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013201.1 | NP_001013219.1 | 1 - 2 | 153 | 98 | |
XM_006233033.2 | XP_006233095.1 | 1 - 2 | 201 | 98 | ||
GenBank mRNA | BC082000.1 | - | 1 - 2 | 153 | 98 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC082000.1, NM_001013201.1, XM_006233033.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
