Assay Details
Gene Symbol: Gdap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013201.1 | NP_001013219.1 | 4 - 5 | 688 | 89 | |
XM_006233032.2 | XP_006233094.1 | 5 - 6 | 786 | 89 | ||
XM_006233033.2 | XP_006233095.1 | 4 - 5 | 736 | 89 | ||
GenBank mRNA | BC082000.1 | - | 4 - 5 | 688 | 89 | |
FQ211326.1 | - | 6 - 7 | 869 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
