Assay Details
Gene Symbol: Adamdec1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106046.1 | NP_001099516.1 | 11 - 12 | 1301 | 123 | |
XM_006252229.2 | XP_006252291.1 | 12 - 13 | 1359 | 123 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
