Assay Details
Gene Symbol: Cab39l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011917.1 | NP_001011917.1 | 4 - 5 | 509 | 101 | |
XM_006252098.2 | XP_006252160.1 | 5 - 6 | 613 | 101 | ||
XM_006252099.2 | XP_006252161.1 | 6 - 7 | 697 | 101 | ||
XM_006252100.2 | XP_006252162.1 | 7 - 8 | 893 | 101 | ||
XM_006252101.2 | XP_006252163.1 | 6 - 7 | 809 | 101 | ||
XM_008770742.1 | XP_008768964.1 | 6 - 7 | 706 | 101 | ||
XM_008770743.1 | XP_008768965.1 | 3 - 4 | 1103 | 101 | ||
GenBank mRNA | BC083684.1 | - | 4 - 5 | 509 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
