Assay Details
Gene Symbol: Chrna2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133420.1 | NP_596911.1 | 3 - 4 | 526 | 94 | |
XM_008770735.1 | XP_008768957.1 | 3 - 4 | 368 | 94 | ||
GenBank mRNA | AY574253.1 | - | 2 - 3 | 222 | 94 | |
L10077.1 | - | 3 - 4 | 526 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
