Assay Details
Gene Symbol: Clu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006252094.2 | XP_006252156.1 | 5 - 6 | 998 | 61 | |
GenBank mRNA | AF314657.1 | - | 2 - 3 | 136 | 61 | |
BC061534.1 | - | 5 - 6 | 899 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
