Assay Details
Gene Symbol: Pbk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079937.1 | NP_001073406.1 | 3 - 4 | 404 | 105 | |
XM_006252102.1 | XP_006252164.1 | 3 - 4 | 285 | 105 | ||
XM_006252103.2 | XP_006252165.1 | 3 - 4 | 263 | 105 | ||
XM_006252104.2 | XP_006252166.1 | 3 - 4 | 274 | 105 | ||
XM_006252105.2 | XP_006252167.1 | 3 - 4 | 543 | 105 | ||
XM_006252106.2 | XP_006252168.1 | 3 - 4 | 483 | 105 | ||
GenBank mRNA | BC129099.1 | - | 3 - 4 | 404 | 105 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
