Assay Details
Gene Symbol: LOC102553828 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006229376.2 | XP_006229438.1 | 1 - 2 | 395 | 64 |
Gene Symbol: Spata13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191686.1 | NP_001178615.1 | 11 - 12 | 3730 | 64 | |
XM_006252113.2 | XP_006252175.1 | 11 - 12 | 3797 | 64 | ||
XM_006252114.2 | XP_006252176.1 | 10 - 11 | 4271 | 64 | ||
XM_006252115.2 | XP_006252177.1 | 8 - 9 | 2771 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
