Assay Details
Gene Symbol: Ppp2r2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053999.2 | NP_446451.1 | 8 - 9 | 1280 | 142 | |
XM_008770730.1 | XP_008768952.1 | 8 - 9 | 2087 | 142 | ||
XM_008770731.1 | XP_008768953.1 | 8 - 9 | 2087 | 142 | ||
XM_008770732.1 | XP_008768954.1 | 6 - 7 | 869 | 142 | ||
GenBank mRNA | BC128731.1 | - | 8 - 9 | 1280 | 142 | |
M83297.1 | - | 9 - 10 | 1226 | 142 | ||
M83298.1 | - | 8 - 9 | 1261 | 142 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
