Assay Details
Gene Symbol: Ctsb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022597.2 | NP_072119.2 | 6 - 7 | 663 | 67 | |
GenBank mRNA | BC072490.1 | - | 6 - 7 | 626 | 67 | |
M11305.1 | - | 3 - 4 | 327 | 67 | ||
X82396.1 | - | 6 - 7 | 592 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
