Assay Details
Gene Symbol: Hpgd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024390.2 | NP_077366.2 | 2 - 3 | 233 | 93 | |
GenBank mRNA | BC062399.1 | - | 2 - 3 | 233 | 93 | |
U44750.1 | - | 2 - 3 | 249 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
