Assay Details
Gene Symbol: Hpgd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024390.2 | NP_077366.2 | 3 - 4 | 332 | 159 | |
GenBank mRNA | BC062399.1 | - | 3 - 4 | 332 | 159 | |
U44750.1 | - | 3 - 4 | 348 | 159 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
