Assay Details
Gene Symbol: Cirbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006241012.2 | XP_006241074.1 | 6 - 7 | 2488 | 72 | |
XM_006241013.2 | XP_006241075.2 | 6 - 7 | 2599 | 72 | ||
XM_006241014.2 | XP_006241076.1 | 7 - 8 | 2287 | 72 | ||
XM_006241015.2 | XP_006241077.2 | 7 - 8 | 2398 | 72 | ||
XM_008765132.1 | XP_008763354.1 | 7 - 8 | 893 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
