Assay Details
Gene Symbol: Cirbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031147.2 | NP_112409.2 | 2 - 3 | 274 | 69 | |
XM_006241012.2 | XP_006241074.1 | 2 - 3 | 279 | 69 | ||
XM_006241013.2 | XP_006241075.2 | 2 - 3 | 390 | 69 | ||
XM_006241014.2 | XP_006241076.1 | 2 - 3 | 279 | 69 | ||
XM_006241015.2 | XP_006241077.2 | 2 - 3 | 390 | 69 | ||
XM_008765132.1 | XP_008763354.1 | 2 - 3 | 390 | 69 | ||
GenBank mRNA | AB000362.1 | - | 1 - 2 | 123 | 69 | |
BC069219.1 | - | 2 - 3 | 274 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
