Assay Details
Gene Symbol: Cirbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031147.2 | NP_112409.2 | 6 - 7 | 668 | 65 | |
XM_006241014.2 | XP_006241076.1 | 6 - 7 | 673 | 65 | ||
XM_006241015.2 | XP_006241077.2 | 6 - 7 | 784 | 65 | ||
GenBank mRNA | BC069219.1 | - | 6 - 7 | 668 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
