Assay Details
Gene Symbol: Apba3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031781.1 | NP_113969.1 | 5 - 6 | 1082 | 98 | |
XM_006241018.2 | XP_006241080.1 | 6 - 7 | 1243 | 98 | ||
XM_006241019.2 | XP_006241081.1 | 6 - 7 | 1216 | 98 | ||
XM_006241020.2 | XP_006241082.1 | 6 - 7 | 1241 | 98 | ||
GenBank mRNA | AF029109.1 | - | 5 - 6 | 1082 | 98 | |
BC161842.1 | - | 6 - 7 | 1144 | 98 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
