Assay Details
Gene Symbol: Gpx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039849.2 | NP_001034938.1 | 4 - 5 | 658 | 124 | |
NM_017165.2 | NP_058861.3 | 4 - 5 | 544 | 124 | ||
GenBank mRNA | AB072798.1 | - | 4 - 5 | 658 | 124 | |
AF274028.1 | - | 4 - 5 | 667 | 124 | ||
AY570513.1 | - | 4 - 5 | 646 | 124 | ||
FQ210610.1 | - | 4 - 5 | 452 | 124 | ||
FQ221663.1 | - | 4 - 5 | 474 | 124 | ||
FQ221760.1 | - | 4 - 5 | 452 | 124 | ||
FQ221899.1 | - | 4 - 5 | 452 | 124 | ||
FQ221976.1 | - | 4 - 5 | 455 | 124 | ||
FQ222299.1 | - | 4 - 5 | 452 | 124 | ||
FQ227764.1 | - | 4 - 5 | 611 | 124 | ||
FQ230991.1 | - | 4 - 5 | 440 | 124 | ||
FQ231108.1 | - | 4 - 5 | 510 | 124 | ||
FQ233099.1 | - | 4 - 5 | 452 | 124 | ||
FQ234630.1 | - | 4 - 5 | 452 | 124 | ||
L24896.1 | - | 4 - 5 | 421 | 124 | ||
U37427.1 | - | 4 - 5 | 538 | 124 | ||
X82679.1 | - | 4 - 5 | 431 | 124 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
