Assay Details
Gene Symbol: Gpx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039849.2 | NP_001034938.1 | 6 - 7 | 745 | 76 | |
NM_017165.2 | NP_058861.3 | 6 - 7 | 631 | 76 | ||
GenBank mRNA | AB072798.1 | - | 6 - 7 | 745 | 76 | |
FQ210610.1 | - | 6 - 7 | 539 | 76 | ||
FQ219811.1 | - | 6 - 7 | 539 | 76 | ||
FQ221663.1 | - | 6 - 7 | 561 | 76 | ||
FQ221899.1 | - | 6 - 7 | 539 | 76 | ||
FQ221976.1 | - | 6 - 7 | 542 | 76 | ||
FQ222299.1 | - | 6 - 7 | 539 | 76 | ||
FQ227564.1 | - | 6 - 7 | 527 | 76 | ||
FQ227764.1 | - | 6 - 7 | 698 | 76 | ||
FQ230991.1 | - | 6 - 7 | 527 | 76 | ||
FQ231108.1 | - | 6 - 7 | 597 | 76 | ||
FQ231686.1 | - | 6 - 7 | 539 | 76 | ||
FQ233099.1 | - | 6 - 7 | 539 | 76 | ||
FQ234630.1 | - | 6 - 7 | 539 | 76 | ||
L24896.1 | - | 6 - 7 | 508 | 76 | ||
X82679.1 | - | 6 - 7 | 518 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
