Assay Details
Gene Symbol: Stk11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108069.1 | NP_001101539.1 | 5 - 6 | 1556 | 113 | |
XM_006240910.1 | XP_006240972.1 | 5 - 6 | 1551 | 113 | ||
XM_008765096.1 | XP_008763318.1 | 5 - 6 | 1551 | 113 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
