Assay Details
Gene Symbol: Epb41l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021681.2 | NP_067713.2 | 21 - 22 | 4526 | 127 | |
NM_172090.3 | NP_742087.1 | 18 - 19 | 2493 | 127 | ||
XM_008762381.1 | XP_008760603.1 | 21 - 22 | 4526 | 127 | ||
GenBank mRNA | AB019256.1 | - | 18 - 19 | 2491 | 127 | |
AB019257.1 | - | 21 - 22 | 4526 | 127 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
