Assay Details
Gene Symbol: Rbm39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013207.2 | NP_001013225.1 | 1444 | 111 | ||
XM_008762356.1 | XP_008760578.1 | 1439 | 111 | |||
GenBank mRNA | BC091394.1 | - | 1433 | 111 |
Gene Symbol: LOC100910882 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003749606.3 | XP_003749654.1 | 12 - 13 | 1502 | 111 | |
XM_008762422.1 | XP_008760644.1 | 11 - 12 | 1439 | 111 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
