Assay Details
Gene Symbol: Raly | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011958.1 | NP_001011958.1 | 2 - 3 | 150 | 104 | |
XM_008762314.1 | XP_008760536.1 | 2 - 3 | 540 | 104 | ||
XM_008762316.1 | XP_008760538.1 | 2 - 3 | 218 | 104 | ||
GenBank mRNA | BC087074.1 | - | 2 - 3 | 150 | 104 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC087074.1, NM_001011958.1, XM_008762314.1, XM_008762316.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
