Assay Details
Gene Symbol: Raly | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011958.1 | NP_001011958.1 | 7 - 8 | 1001 | 92 | |
XM_008762314.1 | XP_008760536.1 | 8 - 9 | 1439 | 92 | ||
XM_008762315.1 | XP_008760537.1 | 7 - 8 | 1157 | 92 | ||
XM_008762316.1 | XP_008760538.1 | 7 - 8 | 1069 | 92 | ||
XM_008762317.1 | XP_008760539.1 | 6 - 7 | 1105 | 92 | ||
GenBank mRNA | BC087074.1 | - | 7 - 8 | 1001 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
