Assay Details
Gene Symbol: Cbfa2t2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168542.1 | NP_001162014.1 | 5 - 6 | 806 | 162 | |
XM_006235311.2 | XP_006235373.1 | 5 - 6 | 790 | 162 | ||
XM_006235312.2 | XP_006235374.1 | 6 - 7 | 818 | 162 | ||
XM_008762313.1 | XP_008760535.1 | 5 - 6 | 894 | 162 | ||
GenBank mRNA | FQ222452.1 | - | 5 - 6 | 822 | 162 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
