Assay Details
Gene Symbol: Csnk2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053824.2 | NP_446276.1 | 9 - 10 | 801 | 115 | |
XM_006235220.2 | XP_006235282.1 | 8 - 9 | 861 | 115 | ||
XM_006235221.2 | XP_006235283.1 | 8 - 9 | 902 | 115 | ||
GenBank mRNA | BC091130.1 | - | 9 - 10 | 801 | 115 | |
L15618.1 | - | 1418 | 115 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
