Assay Details
Gene Symbol: Hm13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006235269.1 | XP_006235331.1 | 11 - 12 | 1158 | 71 | |
GenBank mRNA | FQ218565.1 | - | 4 - 5 | 308 | 71 | |
FQ220534.1 | - | 4 - 5 | 308 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
