Assay Details
Gene Symbol: Sla2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172118.1 | NP_001165589.1 | 1 - 2 | 103 | 105 | |
XM_008762374.1 | XP_008760596.1 | 3 - 4 | 1780 | 105 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
