Assay Details
Gene Symbol: Amfr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008772340.1 | XP_008770562.1 | 4 - 5 | 685 | 123 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
