Assay Details
Gene Symbol: Nudt21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039004.1 | NP_001034093.1 | 3 - 4 | 466 | 91 | |
XM_006255132.2 | XP_006255194.1 | 3 - 4 | 1099 | 91 | ||
GenBank mRNA | BC098748.1 | - | 3 - 4 | 466 | 91 | |
BC168149.1 | - | 3 - 4 | 1105 | 91 | ||
FQ213930.1 | - | 3 - 4 | 509 | 91 | ||
FQ229498.1 | - | 3 - 4 | 507 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
