Assay Details
Gene Symbol: Gnao1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017327.1 | NP_059023.1 | 6 - 7 | 1429 | 129 | |
XM_008772320.1 | XP_008770542.1 | 6 - 7 | 1438 | 129 | ||
XM_008772322.1 | XP_008770544.1 | 5 - 6 | 4047 | 129 | ||
GenBank mRNA | M12671.1 | - | 5 - 6 | 593 | 129 | |
M17526.1 | - | 6 - 7 | 1429 | 129 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
