Assay Details
Gene Symbol: Cdh11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053392.1 | NP_445844.1 | 9 - 10 | 1866 | 61 | |
XM_006255059.2 | XP_006255121.1 | 9 - 10 | 3946 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
