Assay Details
Gene Symbol: Cdh11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053392.1 | NP_445844.1 | 10 - 11 | 2004 | 128 | |
XM_006255059.2 | XP_006255121.1 | 10 - 11 | 4084 | 128 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
