Assay Details
Gene Symbol: Mmp15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106168.2 | NP_001099638.1 | 5 - 6 | 1444 | 68 | |
GenBank mRNA | CB545319.1 | - | 2 - 3 | 441 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
