Assay Details
Gene Symbol: Ecd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008770584.1 | XP_008768806.1 | 10 - 11 | 1561 | 68 | |
XM_008770585.1 | XP_008768807.1 | 11 - 12 | 1420 | 68 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
