Assay Details
Gene Symbol: Ecd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008770584.1 | XP_008768806.1 | 4 - 5 | 735 | 65 | |
XM_008770585.1 | XP_008768807.1 | 5 - 6 | 594 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
