Assay Details
Gene Symbol: Adk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012895.3 | NP_037027.2 | 6 - 7 | 564 | 84 | |
XM_006251631.1 | XP_006251693.1 | 6 - 7 | 678 | 84 | ||
GenBank mRNA | BC081712.1 | - | 6 - 7 | 564 | 84 | |
FQ218369.1 | - | 2 - 3 | 227 | 84 | ||
FQ225093.1 | - | 2 - 3 | 235 | 84 | ||
U90340.1 | - | 6 - 7 | 567 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
