Assay Details
Gene Symbol: Tbpl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127201.1 | NP_001120673.1 | 1 - 2 | 212 | 99 | |
XM_006227721.2 | XP_006227783.1 | 1 - 2 | 245 | 99 | ||
GenBank mRNA | BF414163.1 | - | 1 - 2 | 212 | 99 | |
FQ234587.1 | - | 1 - 2 | 230 | 99 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_001127201.1, XM_006227721.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
