Assay Details
Gene Symbol: Trmt11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198051.1 | NP_932168.1 | 10 - 11 | 1036 | 140 | |
XM_006227748.2 | XP_006227810.1 | 10 - 11 | 1050 | 140 | ||
XM_006227749.2 | XP_006227811.1 | 10 - 11 | 1567 | 140 | ||
XM_006227750.2 | XP_006227812.1 | 10 - 11 | 1049 | 140 | ||
XM_006227751.2 | XP_006227813.1 | 10 - 11 | 1048 | 140 | ||
XM_006227752.2 | XP_006227814.1 | 10 - 11 | 1048 | 140 | ||
XM_008758673.1 | XP_008756895.1 | 10 - 11 | 1710 | 140 | ||
GenBank mRNA | AF532978.1 | - | 10 - 11 | 1036 | 140 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
