Assay Details
Gene Symbol: Trmt11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198051.1 | NP_932168.1 | 9 - 10 | 969 | 77 | |
XM_006227748.2 | XP_006227810.1 | 9 - 10 | 983 | 77 | ||
XM_006227749.2 | XP_006227811.1 | 9 - 10 | 1500 | 77 | ||
XM_006227750.2 | XP_006227812.1 | 9 - 10 | 982 | 77 | ||
XM_006227751.2 | XP_006227813.1 | 9 - 10 | 981 | 77 | ||
XM_006227752.2 | XP_006227814.1 | 9 - 10 | 981 | 77 | ||
XM_008758673.1 | XP_008756895.1 | 9 - 10 | 1643 | 77 | ||
GenBank mRNA | AF532978.1 | - | 9 - 10 | 969 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
